Order Entry
Export
ContactUsLinkComponent
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: Ubi-1]
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: Ubi-1]
Supplier:  Antibodies.com
New Product
Certificates
undefined
Anti-Ubiquitin Mouse Monoclonal Antibody [clone: Ubi-1]
Supplier:  Antibodies.com
New Product

Specifications

  • Catalog No:
  • ANTIA85456-100
  • ANTIA85456-50
  • Antigen symbol:
  • Ubiquitin
  • Ubiquitin
  • Antigen name:
  • Epididymis secretory protein Li 50
  • Epididymis secretory protein Li 50
  • Antigen synonyms:
  • UBA 80|RPS27A|UBA52|FLJ25987|MGC8385|RPS 27A|Polyubiquitin B|HEL S 50|UBA 52
  • UBA 80|RPS27A|UBA52|FLJ25987|MGC8385|RPS 27A|Polyubiquitin B|HEL S 50|UBA 52
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • Ubi-1
  • Ubi-1
  • Reactivity:
  • Danio, Drosophila, Pig, Horse, Monkey, Chicken, C. elegans, Human, Bovine, Rat, Mouse
  • Danio, Drosophila, Pig, Horse, Monkey, Chicken, C. elegans, Human, Bovine, Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID# P0CG48, P0CG47, P62987, P62979
  • UniprotID# P0CG48, P0CG47, P62987, P62979
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Purified bovine erythrocyte Ubiquitin, coupled to KLH with glutaraldehyde
  • Purified bovine erythrocyte Ubiquitin, coupled to KLH with glutaraldehyde
  • Form:
  • Liquid
  • Liquid
  • Molecular weight:
  • 8.5 kDa (monoubiquitin)
  • 8.5 kDa (monoubiquitin)
  • Purification:
  • Immunogen affinity purification.
  • Immunogen affinity purification.
  • Concentration:
  • 1 mg/ml
  • 1 mg/ml
  • Storage buffer:
  • Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
  • Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
  • Storage temperature:
  • Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
  • Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at +4 °C
  • Shipped on blue ice at +4 °C
  • Tested applications:
  • ICC/IF, IHC
  • ICC/IF, IHC

Specifications