Anti-Ubiquitin Rabbit Polyclonal Antibody
Supplier: Antibodies.com
New Product
Certificates
About this item
Rabbit polyclonal antibody to Ubiquitin for WB, ICC/IF and IHC with samples derived from Human, Rat and Mouse.
- Validated Applications: WB, ICC/IF, IHC
- Recommended Dilutions: WB: 1:5000 to 1:10000, ICC/IF: 1:500 to 1:1000, IHC: 1:500 to 1:100
Type: Primary
Antigen: Ubiquitin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat;Mouse
Specifications
- Catalog No:
- ANTIA85455-50
- ANTIA85455-100
- Antigen symbol:
- Ubiquitin
- Ubiquitin
- Antigen name:
- Epididymis secretory protein Li 50
- Epididymis secretory protein Li 50
- Antigen synonyms:
- UBA 80|RPS27A|UBA52|FLJ25987|MGC8385|RPS 27A|Polyubiquitin B|HEL S 50|UBA 52
- UBA 80|RPS27A|UBA52|FLJ25987|MGC8385|RPS 27A|Polyubiquitin B|HEL S 50|UBA 52
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# P0CG48, P0CG47, P62987, P62979
- UniprotID# P0CG48, P0CG47, P62987, P62979
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Bovine blood derived Ubiquitin coupled to keyhole limpet hemocyanin with glutaraldehyde
- Bovine blood derived Ubiquitin coupled to keyhole limpet hemocyanin with glutaraldehyde
- Form:
- Liquid
- Liquid
- Molecular weight:
- 8.5 kDa (monoubiquitin)
- 8.5 kDa (monoubiquitin)
- Storage buffer:
- Supplied as an aliquot of serum with 5 mM Sodium azide
- Supplied as an aliquot of serum with 5 mM Sodium azide
- Storage temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- ICC/IF, IHC
- ICC/IF, IHC