Order Entry
Export
ContactUsLinkComponent
 
Anti-ICAM1 Rabbit Polyclonal Antibody
  ANTIA9725-100
New Product
undefined
Anti-ICAM1 Rabbit Polyclonal Antibody
  ANTIA9725-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Antigen identified by monoclonal
  • Antigen symbol:
    ICAM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05362
  • Antigen synonyms:
    Human rhinovirus receptor|CD54 antigen|Cell surface glycoprotein P3.58|BB2|CD54|CD_antigen=CD54|BB 2|CD 54|ICAM 1
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,05% Proclin 300
  • Molecular weight:
    89 kDa/92 kDa
  • Sequence:
    PLPSWQPVGKNLTLRCQVEGGAPRANLTVVLLRGEKELKREPAVGEPAEVTTTVLVRRDHHGANFSCRTELDLRPQGLELFENTSAPYQLQTFVLPATPPQLVSPRVLEVDTQGTVVCSLDGLFPVSEAQVHLALGDQRLNPTVTYGNDSFSAKASVSVTAEDEGTQRLTCAVILGNQSQETLQTVTIYSFPAPNVILTKP
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 120-320 of human ICAM-1/CD54 (NP_000192.2)
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Affinity purification.
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to ICAM1 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC, ICC/IF
  • Recommended Dilutions: WB: 1:1000 to 1:5000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:20

Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat