Order Entry
Export
Orders LinkContactUsLinkComponent
Anti-GLRB Rabbit Polyclonal Antibody

Anti-GLRB Rabbit Polyclonal Antibody

Plastic vial
Catalog # ANTIA98124-100
Supplier:   Antibodies.com
Anti-GLRB Rabbit Polyclonal Antibody
Catalog # ANTIA98124-100
Supplier:  Antibodies.com
Anti-GLRB Rabbit Polyclonal Antibody

Plastic vial

Price
495,00 €
Log in for availability
1 * 100 µG
Product Details & Documents

Rabbit polyclonal antibody to GLRB for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000

Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.

Type: Primary
Antigen: GLRB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Specifications
  • Seq:
    1286
  • Pack type:
    Plastic vial
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Antibody type:
    Primary
  • Form:
    Liquid
  • Isotype:
    IgG
  • Storage temperature:
    Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Host:
    Rabbit
  • Molecular weight:
    56 kDa
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Antigen name:
    GLRB
  • Cat. No.:
    ANTIA98124-100
  • Reactivity:
    Human|Mouse|Rat
  • Gene ID:
    UniprotID# P48167
  • Antigen synonyms:
    GLRB
  • Immunogen:
    Synthetic peptide derived from human GLRB (amino acids 211 - 260)
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    GLRB
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • ELISA:
    Yes