Order Entry
Export
ContactUsLinkComponent
Anti-ERGI3 Rabbit Polyclonal Antibody
Anti-ERGI3 Rabbit Polyclonal Antibody
Catalog # ANTIA97846-100
Supplier:  Antibodies.com
New Product
undefined
Anti-ERGI3 Rabbit Polyclonal Antibody
Catalog # ANTIA97846-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    2310015B14Rik
  • Antigen symbol:
    ERGI3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9Y282
  • Antigen synonyms:
    Endoplasmic reticulum-Golgi intermediate compartment protein 3|D2Ucla1|endoplasmic reticulum localized protein ERp43|C20orf47|AV318804|Endoplasmic reticulum Golgi intermediate compartment protein 3|DKFZp547A2190|dJ477O4.2|CGI 54
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    43 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human ERGI3 (amino acids 321-370)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to ERGI3 for WB, IF and ELISA with samples derived from Human and Mouse.

  • Validated Applications: WB, IF, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:20000

Type: Primary
Antigen: ERGI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse