Anti-PIP5K3 Rabbit Polyclonal Antibody
Catalog # ANTIA97838-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:1 phosphatidylinositol 3 phosphate 5 kinase
- Antigen symbol:PIP5K3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q9Y2I7
- Antigen synonyms:FYVE finger containing phosphoinositide kinase|1-phosphatidylinositol 3-phosphate 5-kinase|FYVE finger-containing phosphoinositide kinase|CFD|FAB1|FYV1_HUMAN|FAB1, S. cerevisiae, homolog of|1 phosphatidylinositol 4 phosphate 5 kinase|epididymis luminal protein 37
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:237 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human PIP5K (amino acids 71-120)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to PIP5K for WB, IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:5000
Type: Primary
Antigen: PIP5K3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse