Anti-CD57 Rabbit Polyclonal Antibody
Catalog # ANTIA97790-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:3-glucuronyltransferase
- Antigen symbol:CD57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# Q9P2W7
- Antigen synonyms:Beta 1 3 glucuronyltransferase 1|3-glucuronyltransferase 1|CD57|Beta-1|CD 57|B3gat1|CD57 antigen|Beta-1,3-glucuronyltransferase 1|B3GA1_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:38 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human CD57 (amino acids 35-84)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to CD57 for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: CD57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat