Anti-GTF2A1 Rabbit Polyclonal Antibody
Catalog # ANTIA97166-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:General transcription factor IIA 1 19/37kDa
- Antigen symbol:GTF2A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P52655
- Antigen synonyms:TF2AA_HUMAN|Glucose regulated protein 58kD pseudogene|General transcription factor IIA subunit 1|TFIIA 42|TFIIA alpha p55, isoform 1|General transcription factor IIA1|TFIIA alpha p55|TFIIA p19 subunit|GTF2A1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:41 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human TF2A1 (amino acids 281-330)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to TF2A1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000
Type: Primary
Antigen: GTF2A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat