Anti-PIP5K3 Rabbit Polyclonal Antibody
ANTIA97008-100
New Product
- Antibody type:Primary
- Antigen name:1 phosphatidylinositol 3 phosphate 5 kinase
- Antigen symbol:PIP5K3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# Q9Y2I7
- Antigen synonyms:FYVE finger containing phosphoinositide kinase|1-phosphatidylinositol 3-phosphate 5-kinase|FYVE finger-containing phosphoinositide kinase|CFD|FAB1|FYV1_HUMAN|FAB1, S. cerevisiae, homolog of|1 phosphatidylinositol 4 phosphate 5 kinase|epididymis luminal protein 37
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:237 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human PIP5K (amino acids 273-322)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to PIP5K for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:1000
Type: Primary
Antigen: PIP5K3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat