Anti-Hsc70 Rabbit Polyclonal Antibody
ANTIA96860-100
New Product
- Antibody type:Primary
- Antigen name:2410008N15Rik
- Antigen symbol:Hsc70
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P11142
- Antigen synonyms:Epididymis luminal protein 33|Heat shock 70kDa protein 8|Heat shock cognate protein 54|Epididymis secretory sperm binding protein Li 72p|Heat shock 70kD protein 8|Heat shock 70 kDa protein 8|Heat shock 70kD protein 10|Heat shock cognate 71 kDa protein|Constitutive heat shock protein 70
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:70 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human HSC 70 (amino acids 202-251)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to HSC 70 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Hsc70
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat