Order Entry
Export
ContactUsLinkComponent
 
Anti-JNK3 Rabbit Polyclonal Antibody
Catalog # ANTIA96834-100
Supplier:  Antibodies.com
New Product
undefined
Anti-JNK3 Rabbit Polyclonal Antibody
Catalog # ANTIA96834-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    c Jun kinase 3
  • Antigen symbol:
    JNK3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P53779
  • Antigen synonyms:
    JNK3|JNK3A|FLJ33785|JNK3 alpha protein kinase|cJun N terminal kinase 3|MAP kinase 10|FLJ12099|MAP kinase|c-Jun N-terminal kinase 3
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    52 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human MAPK10 (amino acids 361-410)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to MAPK10 for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:20000

Type: Primary
Antigen: JNK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat