Order Entry
Export
ContactUsLinkComponent
 
Anti-PLXNC1 Rabbit Polyclonal Antibody
Catalog # ANTIA96790-100
Supplier:  Antibodies.com
New Product
undefined
Anti-PLXNC1 Rabbit Polyclonal Antibody
Catalog # ANTIA96790-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD232
  • Antigen symbol:
    PLXNC1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O60486
  • Antigen synonyms:
    PLXC1_HUMAN|PLXNC1|Plexin-C1|Virus-encoded semaphorin protein receptor
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    175 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from the Internal region of human PLXNC1 (amino acids 811-860)
  • Purification:
    The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to PLXNC1 for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: PLXNC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat