Anti-SCAMP1 Rabbit Polyclonal Antibody
Catalog # ANTIA96746-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:SCAM1_HUMAN
- Antigen symbol:SCAMP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O15126
- Antigen synonyms:SCAMP 1|SCAMP1|Secretory carrier associated membrane protein 1|SCAMP 37|SCAMP|SCAMP37|Secretory carrier membrane protein|Secretory carrier membrane protein 1|Secretory carrier-associated membrane protein 1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:37 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human SCAMP1 (amino acids 254-303)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to SCAMP1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: SCAMP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat