Anti-Cytokeratin 7+17 Mouse Monoclonal Antibody [clone: ABT-CK17]
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Mouse monoclonal (ABT-CK17) antibody to Cytokeratin 17 for IHC and WB with samples derived from Human.
- Validated Applications: IHC, WB
Type: Primary
Antigen: Cytokeratin 7+17
Clonality: Monoclonal
Clone: ABT-CK17
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ANTIA95694-100
- ANTIA95694-50
- Antigen symbol:
- Cytokeratin 7+17
- Cytokeratin 7+17
- Antigen name:
- CK17
- CK17
- Antigen synonyms:
- CK7|KRT7|Cytokeratin 17|Cytokeratin 7|KRT17|Cytokeratin 7+17
- CK7|KRT7|Cytokeratin 17|Cytokeratin 7|KRT17|Cytokeratin 7+17
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-CK17
- ABT-CK17
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# Q04695
- UniprotID# Q04695
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human Cytokeratin 17
- Synthetic peptide derived from human Cytokeratin 17
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage buffer:
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- IHC
- IHC