Anti-Cytokeratin 8 + 18 + 19 Mouse Monoclonal Antibody [clone: ABT-CK19]
Supplier: ANTIBODIES.COM
New Product
Certificates
About this item
Mouse monoclonal (ABT-CK19) antibody to Cytokeratin 19 for IHC and WB with samples derived from Human.
- Validated Applications: IHC, WB
Type: Primary
Antigen: Cytokeratin 8 + 18 + 19
Clonality: Monoclonal
Clone: ABT-CK19
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ANTIA95692-100
- ANTIA95692-50
- Antigen symbol:
- Cytokeratin 8 + 18 + 19
- Cytokeratin 8 + 18 + 19
- Antigen name:
- CK18
- CK18
- Antigen synonyms:
- keratin 19|Keratin type I cytoskeletal 19|Cytokeratin 8 + 18 + 19|CK19|keratin 18|Keratin type I 40kDa|keratin 8|Keratin type I cytoskeletal 18|CK8
- keratin 19|Keratin type I cytoskeletal 19|Cytokeratin 8 + 18 + 19|CK19|keratin 18|Keratin type I 40kDa|keratin 8|Keratin type I cytoskeletal 18|CK8
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-CK19
- ABT-CK19
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P08727
- UniprotID# P08727
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human Cytokeratin 19
- Synthetic peptide derived from human Cytokeratin 19
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage buffer:
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Shipping temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested applications:
- IHC
- IHC