Order Entry
Export
ContactUsLinkComponent
Anti-Ku80 Mouse Monoclonal Antibody [Clone: MEM-54]
Anti-Ku80 Mouse Monoclonal Antibody [Clone: MEM-54]
Catalog # ANTIA85990-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Ku80 Mouse Monoclonal Antibody [Clone: MEM-54]
Catalog # ANTIA85990-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    86 kDa subunit of Ku antigen
  • Antigen symbol:
    Ku80
  • Clonality:
    Monoclonal
  • Clone:
    MEM-54
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13010
  • Antigen synonyms:
    ATP dependent DNA helicase 2 subunit 2|CTC85|CTC box binding factor 85 kDa|ATP-dependent DNA helicase 2 subunit 2|ATP dependent DNA helicase II 86 Kd subunit|ATP dependent DNA helicase II|ATP-dependent DNA helicase II 80 kDa subunit|CTC box-binding factor 85 kDa subunit|ATP dependent DNA helicase II 80 kDa subunit
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human whole T-lymphocytes
  • Tested applications:
    ICC
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [MEM-54] antibody to Ku80 for ICC and IP with samples derived from human.

  • Validated applications: ICC, IP
  • Recommended dilutions: ICC: 2 µg/ml

Type: Primary
Antigen: Ku80
Clonality: Monoclonal
Clone: MEM-54
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human