Order Entry
Export
ContactUsLinkComponent
Anti-CD59 Mouse Monoclonal Antibody [Clone: MEM-43]
Anti-CD59 Mouse Monoclonal Antibody [Clone: MEM-43]
Catalog # ANTIA85989-100
Supplier:  ANTIBODIES.COM
New Product
Anti-CD59 Mouse Monoclonal Antibody [Clone: MEM-43]
Catalog # ANTIA85989-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD59 antigen complement regulatory protein
  • Antigen symbol:
    CD59
  • Clonality:
    Monoclonal
  • Clone:
    MEM-43
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13987
  • Antigen synonyms:
    CD59 antigen|1F5|CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344)|CD 59|CD59|16.3A5|20 kDa homologous restriction factor|CD59 antigen p18 20|1F5 antigen
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Thymocytes and T lymphocytes
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [MEM-43] antibody to CD59 for flow cytometry, IP and IHC-P with samples derived from human.

  • Validated applications: Flow cytometry, IP, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/ml, IHC-P: 10 µg/ml

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MEM-43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human