Order Entry
Export
ContactUsLinkComponent
Anti-CD56 Mouse Monoclonal Antibody [Clone: MEM-188]
Anti-CD56 Mouse Monoclonal Antibody [Clone: MEM-188]
Catalog # ANTIA85988-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD56 Mouse Monoclonal Antibody [Clone: MEM-188]
Catalog # ANTIA85988-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    antigen MSK39 identified by monoclonal
  • Antigen symbol:
    CD56
  • Clonality:
    Monoclonal
  • Clone:
    MEM-188
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13591
  • Antigen synonyms:
    NCAM 1|NCAM|N-CAM-1|MSK39|CD56|antigen recognized by monoclonal|cell adhesion molecule, neural, 1|MSK 39|NCAM C
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    KG-1 human acute myelogenous leukemia cell line
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [MEM-188] antibody to CD56 for flow cytometry, IP and IHC-P with samples derived from human and non-human primates.

  • Validated applications: Flow cytometry, IP, IHC-P
  • Recommended dilutions: Flow cytometry: 3 µg/ml, IHC-P: 1:50

Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: MEM-188
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Non-Human Primates