Order Entry
Export
ContactUsLinkComponent
 
Anti-CD56 Mouse Monoclonal Antibody [Clone: MEM-188] (FITC (Fluorescein Isothiocyanate))
  ANTIA85984-100
New Product
undefined
Anti-CD56 Mouse Monoclonal Antibody [Clone: MEM-188] (FITC (Fluorescein Isothiocyanate))
  ANTIA85984-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    antigen MSK39 identified by monoclonal
  • Antigen symbol:
    CD56
  • Clonality:
    Monoclonal
  • Clone:
    MEM-188
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2a
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13591
  • Antigen synonyms:
    NCAM 1|NCAM|N-CAM-1|MSK39|CD56|antigen recognized by monoclonal|cell adhesion molecule, neural, 1|MSK 39|NCAM C
  • Storage buffer:
    Supplied in phosphate buffered saline with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    KG-1 human acute myelogenous leukemia cell line
  • Purification:
    This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

 

 

Mouse monoclonal [MEM-188] antibody to CD56 (FITC) for flow cytometry with samples derived from human and non-human primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: MEM-188
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Non-Human Primates