Anti-CD56 Mouse Monoclonal Antibody [Clone: MEM-188] (PE (Phycoerythrin))
Catalog # ANTIA85971-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:antigen MSK39 identified by monoclonal
- Antigen symbol:CD56
- Clonality:Monoclonal
- Clone:MEM-188
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P13591
- Antigen synonyms:NCAM 1|NCAM|N-CAM-1|MSK39|CD56|antigen recognized by monoclonal|cell adhesion molecule, neural, 1|MSK 39|NCAM C
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:KG-1 human acute myelogenous leukemia cell line
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [MEM-188] antibody to CD56 (PE) for flow cytometry with samples derived from human and non-human primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD56
Clonality: Monoclonal
Clone: MEM-188
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Non-Human Primates