Anti-MZF1 Rabbit Polyclonal Antibody
Catalog # ANTIA8593-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Myeloid zinc finger 1
- Antigen symbol:MZF1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P28698
- Antigen synonyms:Zfp98|Zinc finger and SCAN domain containing protein 6|MZF1|MZF1_HUMAN|MZF1B|Zinc finger protein 42|MZF 1|Zinc finger and SCAN domain-containing protein 6|MZF-1
- Storage buffer:Supplied in phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
- Molecular weight:110 kDa
- Sequence:IQARVQGQRPGSPEEAAALVDGLRREPGGPRRWVTVQVQGQEVLSEKMEPSSFQPLPETEPPTPEPGPKTPPRTMQESPLGLQVKEESEVTEDSDFLESGPLAATQESVPTLLPEEAQRCGTVLDQIFPHSKTGPEGPSWREHPRALWHEEAGGIFSPGFALQLGSISAGPGSVSPHLHVPWDLGMAGLSGQIQSPSREGGFAHALLLPSDLRSEQDPTDEDPCRGVGPALITTRWRSPRGRSRGRPSTGG
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 100-350 of human MZF1 (NP_003413.2)
- Purification:Affinity purification.
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to MZF1 for WB with samples derived from human, mouse and rat.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:200
Type: Primary
Antigen: MZF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat