Order Entry
Export
ContactUsLinkComponent
Anti-KIR2DL4 Mouse Monoclonal Antibody [Clone: mAb#33] (APC (Allophycocyanin))
Anti-KIR2DL4 Mouse Monoclonal Antibody [Clone: mAb#33] (APC (Allophycocyanin))
Catalog # ANTIA85915-100
Supplier:  Antibodies.com
New Product
undefined
Anti-KIR2DL4 Mouse Monoclonal Antibody [Clone: mAb#33] (APC (Allophycocyanin))
Catalog # ANTIA85915-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD158 antigen like family member D
  • Antigen symbol:
    KIR2DL4
  • Clonality:
    Monoclonal
  • Clone:
    mAb#33
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Antigen synonyms:
    CD158d|KI2L4_HUMAN|CD158 antigen-like family member D|CD_antigen=CD158d|Killer cell inhibitory receptor 103AS|Killer cell immunoglobulin like receptor 2DL4|Killer cell immunoglobulin-like receptor 2DL4|Killer cell immunoglobulin like receptor, two domains, long cytoplasmic tail, 4|G9P
  • Storage buffer:
    Supplied in phosphate buffered saline with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    NK3.3 cells and KIR2DL4-Ig fusion protein
  • Purification:
    This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [mAb#33] antibody to KIR2DL4 (APC) for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: KIR2DL4
Clonality: Monoclonal
Clone: mAb#33
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human