Anti-CD206 Mouse Monoclonal Antibody [Clone: 15-2]
Catalog # ANTIA85837-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:bA541I19.1
- Antigen symbol:CD206
- Clonality:Monoclonal
- Clone:15-2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P22897
- Antigen synonyms:CD206|CD206 antigen|C type lectin domain family 13 member D|Macrophage mannose receptor 1|Macrophage mannose receptor|CLEC13D|CD 206|C-type lectin domain family 13 member D|CLEC13DL
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, without sodium azide (0,2 µm filter sterilised). Endotoxin level is less than 0,01 EU/µg of the protein, as determined by the LAL test
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified human mannose receptor
- Tested applications:IHC-Fr, ICC, Functional studies
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [15-2] antibody to CD206 for flow cytometry, IP, WB, IHC-Fr, ICC and functional studies with samples derived from human.
- Validated applications: Flow cytometry, IP, WB, IHC-Fr, ICC, functional studies
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: CD206
Clonality: Monoclonal
Clone: 15-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human