Order Entry
Export
ContactUsLinkComponent
Anti-CD52 Mouse Monoclonal Antibody [Clone: HI186] (PE (Phycoerythrin))
Anti-CD52 Mouse Monoclonal Antibody [Clone: HI186] (PE (Phycoerythrin))
Catalog # ANTIA85764-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD52 Mouse Monoclonal Antibody [Clone: HI186] (PE (Phycoerythrin))
Catalog # ANTIA85764-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cambridge pathology 1 antigen
  • Antigen symbol:
    CD52
  • Clonality:
    Monoclonal
  • Clone:
    HI186
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P31358
  • Antigen synonyms:
    CD 52|CD52 antigen|CAMPATH 1|CD52_HUMAN|CAMPATH-1 antigen|CD52|CDw52|CD52 molecule|CAMPATH 1 antigen
  • Storage buffer:
    Supplied in phosphate buffered saline with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human tonsil
  • Purification:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [HI186] antibody to CD52 (PE) for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD52
Clonality: Monoclonal
Clone: HI186
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human