Anti-CD52 Mouse Monoclonal Antibody [Clone: HI186] (APC (Allophycocyanin))
Catalog # ANTIA85761-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cambridge pathology 1 antigen
- Antigen symbol:CD52
- Clonality:Monoclonal
- Clone:HI186
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P31358
- Antigen synonyms:CD 52|CD52 antigen|CAMPATH 1|CD52_HUMAN|CAMPATH-1 antigen|CD52|CDw52|CD52 molecule|CAMPATH 1 antigen
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human tonsil
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [HI186] antibody to CD52 (APC) for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD52
Clonality: Monoclonal
Clone: HI186
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human