Order Entry
Export
ContactUsLinkComponent
Anti-CD14 Mouse Monoclonal Antibody [Clone: MEM-18]
Anti-CD14 Mouse Monoclonal Antibody [Clone: MEM-18]
  ANTIA85711-100
New Product
Anti-CD14 Mouse Monoclonal Antibody [Clone: MEM-18]
  ANTIA85711-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CD 14
  • Antigen symbol:
    CD14
  • Clonality:
    Monoclonal
  • Clone:
    MEM-18
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Non-human primates,
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P08571
  • Antigen synonyms:
    Monocyte differentiation antigen CD14 urinary form|Mo2|CD_antigen=CD14|CD14 antigen|CD14 molecule|Monocyte differentiation antigen CD14|CD14_HUMAN|CD14|LPS-R
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, without sodium azide (0,2 µm filter sterilised). Endotoxin level is less than 0,01 EU/µg of the protein, as determined by the LAL test
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A crude mixture of human urinary proteins, precipitated by ammonium sulphate, from the urine of a patient suffering from proteinuria
  • Tested applications:
    Functional studies
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [MEM-18] antibody to CD14 for flow cytometry, IP, WB, ELISA and functional studies with samples derived from human and non-human primates.

  • Validated applications: Flow cytometry, IP, WB, ELISA, functional studies
  • Recommended dilutions: Flow cytometry: 4 µg/ml

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: MEM-18
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates