Anti-CD14 Mouse Monoclonal Antibody [Clone: MEM-18] (Biotin)
Catalog # ANTIA85706-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 14
- Antigen symbol:CD14
- Clonality:Monoclonal
- Clone:MEM-18
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P08571
- Antigen synonyms:Monocyte differentiation antigen CD14 urinary form|Mo2|CD_antigen=CD14|CD14 antigen|CD14 molecule|Monocyte differentiation antigen CD14|CD14_HUMAN|CD14|LPS-R
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Store at 2...8 °C. Do not freeze.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A crude mixture of human urinary proteins, precipitated by ammonium sulphate, from the urine of a patient suffering from proteinuria
- Purification:This antibody is conjugated with Biotin-LC-NHS under optimum conditions. The reagent is free of unconjugated Biotin.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MEM-18] antibody to CD14 (Biotin) for flow cytometry, IP, WB and ELISA with samples derived from human and non-human primates.
- Validated applications: Flow cytometry, IP, WB, ELISA
- Recommended dilutions: Flow cytometry: 2 to 4 µg/ml
Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: MEM-18
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates