Anti-CD47 Mouse Monoclonal Antibody [Clone: MEM-122]
Catalog # ANTIA85665-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen identified by monoclonal
- Antigen symbol:CD47
- Clonality:Monoclonal
- Clone:MEM-122
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgM
- Reactivity:Pig,Non-human primates,Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q08722
- Antigen synonyms:IAP|CD47_HUMAN|CD47|CD 47|CD47 antigen|CD47 molecule|Antigenic surface determinant protein OA3|CD47 glycoprotein|CD47 antigen (Rh-related antigen, integrin-associated signal transducer)
- Storage buffer:Supplied in tris buffered saline, pH 8,0, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:COS-7 (African Green Monkey) cells
- Tested applications:IHC-Fr
- Purification:Purified by precipitation and chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [MEM-122] antibody to CD47 for flow cytometry, WB and IHC-Fr with samples derived from human, porcine and non-human primates.
- Validated applications: Flow cytometry, WB, IHC-Fr
- Recommended dilutions: Flow cytometry: 2 µg/ml
Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: MEM-122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Porcine, Non-Human Primates