Anti-CD34 Mouse Monoclonal Antibody [Clone: QBEnd-10]
Catalog # ANTIA85659-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD34
- Clonality:Monoclonal
- Clone:QBEnd-10
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P28906
- Antigen synonyms:CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, without sodium azide (0,2 µm filter sterilised)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human endothelial vesicles
- Tested applications:IHC-P, IHC-Fr, Functional studies
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [QBEnd-10] antibody to CD34 for flow cytometry, IP, WB, IHC-P, IHC-Fr and functional studies with samples derived from human and non-human primates.
- Validated applications: Flow cytometry, IP, WB, IHC-P, IHC-Fr, functional studies
- Recommended dilutions: Flow cytometry: 5 µg/ml, IHC-P: 2 to 8 µg/ml, WB: 1 to 2 µg/ml
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: QBEnd-10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates