Anti-CD34 Mouse Monoclonal Antibody [Clone: 581]
ANTIA85558-100
New Product
- Antibody type:Primary
- Antigen name:CD34
- Clonality:Monoclonal
- Clone:581
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P28906
- Antigen synonyms:CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Unfortunately, this information is considered to be commercially sensitive
- Tested applications:IHC-Fr
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [581] antibody to CD34 for flow cytometry and IHC-Fr with samples derived from human and non-human primates.
- Validated applications: Flow cytometry, IHC-Fr
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 581
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates