Anti-CD34 Mouse Monoclonal Antibody [Clone: 4H11[APG]
ANTIA85518-100
New Product
- Antibody type:Primary
- Antigen name:CD34
- Clonality:Monoclonal
- Clone:4H11[APG
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P28906
- Antigen synonyms:CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Permanent human cell line derived from peripheral leucocytes of a patient suffering from chronic myeloid leukaemia
- Tested applications:IHC-P, ICC
- Purification:Purified by precipitation and chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [4H11[APG]] antibody to CD34 for flow cytometry, WB, IHC-P and ICC with samples derived from human.
- Validated applications: Flow cytometry, WB, IHC-P, ICC
- Recommended dilutions: WB: 1 to 2 µg/ml, Flow cytometry: 2 µg/ml, IHC-P: 10 µg/ml
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11[APG
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human