Order Entry
Export
ContactUsLinkComponent
Anti-CD30 Mouse Monoclonal Antibody [Clone: MEM-268] (PE (Phycoerythrin))
Anti-CD30 Mouse Monoclonal Antibody [Clone: MEM-268] (PE (Phycoerythrin))
Catalog # ANTIA85511-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CD30 Mouse Monoclonal Antibody [Clone: MEM-268] (PE (Phycoerythrin))
Catalog # ANTIA85511-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD 30
  • Antigen symbol:
    CD30
  • Clonality:
    Monoclonal
  • Clone:
    MEM-268
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28908
  • Antigen synonyms:
    CD30|KI1|Cytokine receptor CD30|KI 1|Ki-1 antigen|D1S166E|KI 1 antigen|CD30L receptor|CD30 antigen
  • Storage buffer:
    Supplied in phosphate buffered saline with 15 mM sodium azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Expression vector containing CD30 cDNA (booster suspension of THP-1 cell line)
  • Purification:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-268] antibody to CD30 (PE) for flow cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD30
Clonality: Monoclonal
Clone: MEM-268
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human