Anti-CD30 Mouse Monoclonal Antibody [Clone: MEM-268] (FITC (Fluorescein Isothiocyanate))
Catalog # ANTIA85510-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 30
- Antigen symbol:CD30
- Clonality:Monoclonal
- Clone:MEM-268
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P28908
- Antigen synonyms:CD30|KI1|Cytokine receptor CD30|KI 1|Ki-1 antigen|D1S166E|KI 1 antigen|CD30L receptor|CD30 antigen
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Expression vector containing CD30 cDNA (booster suspension of THP-1 cell line)
- Purification:This antibody is conjugated with FITC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [MEM-268] antibody to CD30 (FITC) for flow cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD30
Clonality: Monoclonal
Clone: MEM-268
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human