Anti-CD22 Mouse Monoclonal Antibody [Clone: MEM-01] (PE (Phycoerythrin))
ANTIA85476-100
New Product
- Antibody type:Primary
- Antigen name:B cell receptor CD22 precursor
- Antigen symbol:CD22
- Clonality:Monoclonal
- Clone:MEM-01
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P20273
- Antigen synonyms:B lymphocyte cell adhesion molecule|CD 22|B-lymphocyte cell adhesion molecule|CD22 molecule|B-cell receptor CD22|BL-CAM|CD22 antigen|BL CAM|BLCAM
- Storage buffer:Supplied in phosphate buffered saline with 15 mM sodium azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Raji Burkitt's lymphoma cell line
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [MEM-01] antibody to CD22 (PE) for flow cytometry with samples derived from human and non-human primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 20 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: MEM-01
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates