Anti-hCG beta Mouse Monoclonal Antibody [Clone: HCG-61]
Catalog # ANTIA85467-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CG beta
- Antigen symbol:hCG beta
- Clonality:Monoclonal
- Clone:HCG-61
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P0DN86
- Antigen synonyms:CGB3|Chorionic gonadotrophin chain beta|CGHB_HUMAN|CG-beta|Choriogonadotropin subunit beta|CGB8|CGB5|CGB|CGB7
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human chorionic gonadotropin
- Tested applications:ICC
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [HCG-61] antibody to hCG beta for ICC and ELISA with samples derived from human.
- Validated applications: ICC, ELISA
Type: Primary
Antigen: hCG beta
Clonality: Monoclonal
Clone: HCG-61
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human