Anti-c-Myc Mouse Monoclonal Antibody [Clone: 9E10] (Biotin)
Catalog # ANTIA85465-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AU016757
- Antigen symbol:c-Myc
- Clonality:Monoclonal
- Clone:9E10
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MRTL|Class E basic helix-loop-helix protein 39|bHLHe39|c Myc|Avian myelocytomatosis viral oncogene homolog|c-Myc|Myc Epitope Tag|Myc protein|Myc proto oncogene protein
- Storage buffer:Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
- Sequence:AEEQKLISEEDLL
- Storage temperature:Store at 2...8 °C. Do not freeze.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the C-terminal region of human c-Myc
- Tested applications:IHC-P
- Purification:This antibody is conjugated with Biotin-LC-NHS under optimum conditions. The reagent is free of unconjugated Biotin.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [9E10] antibody to c-Myc (Biotin) for flow cytometry, IP, WB and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IP, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 5 µg/ml
Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: 9E10
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human