Order Entry
Export
ContactUsLinkComponent
Anti-c-Myc Mouse Monoclonal Antibody [Clone: 9E10]
Anti-c-Myc Mouse Monoclonal Antibody [Clone: 9E10]
  ANTIA85464-100
New Product
Anti-c-Myc Mouse Monoclonal Antibody [Clone: 9E10]
  ANTIA85464-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    AU016757
  • Antigen symbol:
    c-Myc
  • Clonality:
    Monoclonal
  • Clone:
    9E10
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    MRTL|Class E basic helix-loop-helix protein 39|bHLHe39|c Myc|Avian myelocytomatosis viral oncogene homolog|c-Myc|Myc Epitope Tag|Myc protein|Myc proto oncogene protein
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,4, with 15 mM sodium azide
  • Sequence:
    AEEQKLISEEDLL
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to the C-terminal region of human c-Myc
  • Tested applications:
    IHC-P
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [9E10] antibody to c-Myc for flow cytometry, IP, WB and IHC-P with samples derived from human.

  • Validated applications: Flow cytometry, IP, WB, IHC-P
  • Recommended dilutions: IHC-P: 5 to 10 µg/ml, IP: 1 to 5 µg/ml, Flow cytometry: 1 to 4 µg/ml, WB: 0,5 to 2 µg/ml

Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: 9E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human