Anti-c-Myc Mouse Monoclonal Antibody [Clone: 9E10] (HRP (Horseradish Peroxidase))
Catalog # ANTIA85463-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AU016757
- Antigen symbol:c-Myc
- Clonality:Monoclonal
- Clone:9E10
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MRTL|Class E basic helix-loop-helix protein 39|bHLHe39|c Myc|Avian myelocytomatosis viral oncogene homolog|c-Myc|Myc Epitope Tag|Myc protein|Myc proto oncogene protein
- Storage buffer:Supplied in phosphate buffered saline with 0,01% thimerosal
- Sequence:AEEQKLISEEDLL
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to the C-terminal region of human c-Myc
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [9E10] antibody to c-Myc (HRP) for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 1 to 2 µg/ml
Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: 9E10
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human