Order Entry
Export
ContactUsLinkComponent
Anti-GAP43 Chicken Polyclonal Antibody
Anti-GAP43 Chicken Polyclonal Antibody
Supplier:  Antibodies.com
New Product
Certificates
undefined
Anti-GAP43 Chicken Polyclonal Antibody
Supplier:  Antibodies.com
New Product

Specifications

  • Catalog No:
  • ANTIA85292-50
  • ANTIA85292-100
  • Antigen symbol:
  • GAP43
  • GAP43
  • Antigen name:
  • Axonal membrane protein GAP 43
  • Axonal membrane protein GAP 43
  • Antigen synonyms:
  • Axonal membrane protein GAP-43|Growth-associated protein 43|Calmodulin binding protein P 57|Nerve growth related peptide GAP43|Growth Associated Protein 43|Nerve Growth Related Peptide|GAP43|B 50|F1
  • Axonal membrane protein GAP-43|Growth-associated protein 43|Calmodulin binding protein P 57|Nerve growth related peptide GAP43|Growth Associated Protein 43|Nerve Growth Related Peptide|GAP43|B 50|F1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Pig, Horse, Human, Bovine, Rat, Mouse
  • Pig, Horse, Human, Bovine, Rat, Mouse
  • Host:
  • Chicken
  • Chicken
  • Gene ID:
  • UniprotID# P07936
  • UniprotID# P07936
  • Isotype:
  • IgY
  • IgY
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • C-terminal peptide of rodent GAP43, coupled to KLH
  • C-terminal peptide of rodent GAP43, coupled to KLH
  • Sequence:
  • KEDPEADQEHA
  • KEDPEADQEHA
  • Form:
  • Liquid
  • Liquid
  • Molecular weight:
  • ~43 kDa (by SDS-PAGE)
  • ~43 kDa (by SDS-PAGE)
  • Storage buffer:
  • Supplied as an aliquot of IgY preparation with 5 mM sodium azide
  • Supplied as an aliquot of IgY preparation with 5 mM sodium azide
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C
  • Tested applications:
  • ICC/IF, IHC
  • ICC/IF, IHC

Specifications