Anti-MMP14 Goat Polyclonal Antibody
Catalog # ANTIA85106-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Matrix metallopeptidase 14 (membrane inserted)
- Antigen symbol:MMP14
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MMP 14|Membrane type matrix metalloproteinase 1|Membrane-type-1 matrix metalloproteinase|Matrix metalloproteinase-14|MMP X1|Membrane-type matrix metalloproteinase 1|Membrane type 1 matrix metalloproteinase|Matrix metalloproteinase 14|Membrane type 1 metalloprotease
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-ESATPLRFREVP
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human MMP14 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to MMP14 for ELISA, WB and IHC with samples derived from human and mouse.
- Validated applications: ELISA, WB, IH
Type: Primary
Antigen: MMP14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse