Anti-Aspa Goat Polyclonal Antibody
Catalog # ANTIA84937-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:ACY 2
- Antigen symbol:Aspa
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:ACY2_HUMAN|Aminoacylase 2|ACY-2|ASPA|ACY2|ASP|Aminoacylase2|Aspartoacylase|Aminoacylase-2
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-TLSAKSIRSTLH
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Mouse Aspa (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to ASPA for ELISA and WB with samples derived from mouse and rat.
- Validated applications: ELISA, W
Type: Primary
Antigen: Aspa
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse;Rat