Anti-Ascl3 Goat Polyclonal Antibody
Catalog # ANTIA84922-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Achaete scute complex Drosophila homolog like 3
- Antigen symbol:Ascl3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Class A basic helix loop helix protein 42|Achaete scute homolog 3|Achaete-scute homolog 3|ASH 3|Achaete scute complex homolog 3|bHLH transcriptional regulator Sgn 1|HASH3|bHLHa42|ASCL3
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-HLPEDYLEKRLSK
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Mouse Ascl3 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to ASCL3 for ELISA, WB and IHC with samples derived from human and mouse.
- Validated applications: ELISA, WB, IH
Type: Primary
Antigen: Ascl3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse