Order Entry
Export
ContactUsLinkComponent
 
Anti-E2F6 Goat Polyclonal Antibody
  ANTIA84680-100
New Product
undefined
Anti-E2F6 Goat Polyclonal Antibody
  ANTIA84680-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    E2F 6
  • Antigen symbol:
    E2F6
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    MGC111545|E2F binding site modulating activity protein|Transcription factor E2F6|E2F-6|E2F transcription factor 6 isoform 1|EMA|E2F6_HUMAN|E2F6|E2F transcription factor 6
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    C-NPQQSEELLEVSN
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human E2F6 (C terminal)
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Goat polyclonal antibody to E2F6 for ELISA and WB with samples derived from human.

  • Validated applications: ELISA, W

Type: Primary
Antigen: E2F6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human