Anti-IFT57 Goat Polyclonal Antibody
Catalog # ANTIA84663-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CDNA FLJ10147 fis clone HEMBA1003369
- Antigen symbol:IFT57
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Estrogen related receptor beta like protein 1|Estrogen related receptor beta like 1|DERP8|Dermal papilla-derived protein 8|Dermal papilla derived protein 8|DERP 8|Esrrbl 1|ESRRBL1 protein|Esrrbl1
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-EQPQEYDDPNATISN
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human IFT57 (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to IFT57 for ELISA, WB and IHC with samples derived from human and mouse.
- Validated applications: ELISA, WB, IH
Type: Primary
Antigen: IFT57
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse