Anti-Aryl hydrocarbon receptor Goat Polyclonal Antibody
Catalog # ANTIA84652-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Ah receptor
- Antigen symbol:Aryl hydrocarbon receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:Aryl hydrocarbon Receptor|AHR_HUMAN|Aromatic hydrocarbon receptor|AhR|Class E basic helix loop helix protein 76|Aryl hydrocarbon receptor precursor|HGNC:348|Class E basic helix-loop-helix protein 76|bHLHe76
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-PENQKHGLNPQSA
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human AHR (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to Aryl hydrocarbon receptor for ELISA and IHC with samples derived from human.
- Validated applications: ELISA, IH
Type: Primary
Antigen: Aryl hydrocarbon receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human