Order Entry
Export
ContactUsLinkComponent
Anti-UNC5C Goat Polyclonal Antibody
Anti-UNC5C Goat Polyclonal Antibody
Catalog # ANTIA84649-100
Supplier:  Antibodies.com
New Product
undefined
Anti-UNC5C Goat Polyclonal Antibody
Catalog # ANTIA84649-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    homolog of C. elegans transmembrane receptor Unc5
  • Antigen symbol:
    UNC5C
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Form:
    Liquid
  • Antigen synonyms:
    Unc 5 homolog 3|Netrin receptor UNC5C|UNC5H3|Protein unc-5 homolog C|Protein unc 5 homolog C|Protein unc-5 homolog 3|Unc5 (C.elegans homolog) c|UNC5C_HUMAN
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    NCTVSEEPTGID
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human UNC5C (internal region)
  • Tested applications:
    IHC
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Goat polyclonal antibody to UNC5C for ELISA, IF and IHC with samples derived from human and mouse.

  • Validated applications: ELISA, IF, IH

Type: Primary
Antigen: UNC5C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse