Order Entry
Export
ContactUsLinkComponent
Anti-ZRANB1 Goat Polyclonal Antibody
Anti-ZRANB1 Goat Polyclonal Antibody
Catalog # ANTIA84549-100
Supplier:  Antibodies.com
New Product
undefined
Anti-ZRANB1 Goat Polyclonal Antibody
Catalog # ANTIA84549-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    DKFZp762P2216
  • Antigen symbol:
    ZRANB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    TRAF-binding protein domain|Zinc finger Ran-binding domain-containing protein 1|ZRAN1_HUMAN|Ubiquitin thioesterase ZRANB1|hTrabid
  • Storage buffer:
    Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
  • Sequence:
    KWLDRYRQIRPCTS
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    100 µg at 0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide corresponding to Human ZRANB1 (C terminal)
  • Purification:
    Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Goat polyclonal antibody to ZRANB1 for ELISA and WB with samples derived from human.

  • Validated applications: ELISA, W

Type: Primary
Antigen: ZRANB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human