Anti-VPS37C Goat Polyclonal Antibody
Catalog # ANTIA84109-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:5730409F24Rik
- Antigen symbol:VPS37C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:PML39|ESCRT I complex subunit VPS37C|Vacuolar protein sorting 37C|FLJ20847|AU042646|ESCRT-I complex subunit VPS37C|Vacuolar protein sorting 37 homolog C (S. cerevisiae)|RGD1309258|hVps37C
- Storage buffer:Supplied in tris buffered saline, pH 7,30, with 0,02% sodium azide and 0,5% BSA
- Sequence:C-ETLKDKTLQELEELQ
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg at 0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to Human VPS37C (internal region)
- Tested applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Goat polyclonal antibody to VPS37C for ELISA, WB and IHC with samples derived from human.
- Validated applications: ELISA, WB, IH
Type: Primary
Antigen: VPS37C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human