Anti-PKMYT1 Rabbit Polyclonal Antibody
Catalog # ANTIA99870-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:cdc2 inhibitory kinase
- Antigen symbol:PKMYT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q99640
- Antigen synonyms:PKMYT1|Membrane-associated tyrosine- and threonine-specific cdc2-inhibitory kinase|Membrane associated tyrosine and threonine specific cdc2 inhibitory kinase|MYT1|PKMYT 1|Myt1 kinase|FLJ20093|DKFZp547K1610|PMYT1_HUMAN
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:54 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human PKMYT1 (amino acids 49-98)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to MYT1 for WB, IHC and ELISA with samples derived from human.
- Validated applications: WB, IHC, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:20000
Type: Primary
Antigen: PKMYT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human