Anti-TAF1 48 Rabbit Polyclonal Antibody
Catalog # ANTIA99597-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:MGC:17061
- Antigen symbol:TAF1 48
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q15573
- Antigen synonyms:SL1 48kD subunit|TAF1 48|RAFI48|TAF1A_HUMAN|SL1|TAFI48|TATA box binding protein (TBP) associated factor RNA polymerase I, A 48kDa|Taf1a|RNA polymerase I-specific TBP-associated factor 48 kDa
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:52 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TAF1A (amino acids 301-350)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to TAF1A for WB and ELISA with samples derived from human and rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, ELISA: 1:5000
Type: Primary
Antigen: TAF1 48
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat